ATCH1F1C5981
Atlantic Research Chemicals Ltd
Methyl 4-Aminobenzoate hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 4-aminobenzoate hydrochloride
Inchi Code
1S/C8H9NO2.ClH/c1-11-8(10)6-2-4-7(9)5-3-6;/h2-5H,9H2,1H3;1H
InChI key
UMLFQRYMOHDCCK-UHFFFAOYSA-N
Purity
95
Country of Origin
GB
Product Link
https://atlantic-chemicals.com/product/?code=XS0011705
MSDS
https://atlantic-chemicals.com/product/?code=XS0011705
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service