Skip to Content
Merck
All Photos(1)

Key Documents

ATC386058957

Atlantic Research Chemicals Ltd

di-tert-Butyl 2-chloropyridin-3-ylimidodicarbonate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
328.8
MDL number:

Iupac Name

di(tert-butyl) 2-chloro-3-pyridinylimidodicarbonate

Inchi Code

1S/C15H21ClN2O4/c1-14(2,3)21-12(19)18(13(20)22-15(4,5)6)10-8-7-9-17-11(10)16/h7-9H,1-6H3

InChI key

OCIZYSQGQMHVPL-UHFFFAOYSA-N

Purity

90

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=AE002224

MSDS

https://atlantic-chemicals.com/product/?code=AE002224


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library