Skip to Content
Merck
All Photos(1)

Key Documents

ATE458963214

AstaTech, Inc.

TERT-BUTYL 4-(3-IODOPHENYL)PIPERAZINE-1-CARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
388.25
MDL number:

Iupac Name

tert-butyl 4-(3-iodophenyl)piperazine-1-carboxylate

Inchi Code

1S/C15H21IN2O2/c1-15(2,3)20-14(19)18-9-7-17(8-10-18)13-6-4-5-12(16)11-13/h4-6,11H,7-10H2,1-3H3

InChI key

VJXLHWFRDKWCHC-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Shipping Temperature

Normal

Physical Form

Yellow to Brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=72092

Signal Word

Warning

Hazard Statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=21192


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library