Z757993
Q-Tube™ gas purging set
pkg of 1 ea
Synonym(s):
Q-Tube™ gas purging set, pressure reactor, q-tube pressure reactor
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Q Labtech
General description
This set can be operated safely and accurately with high reproducibility with a wide range of gases, including oxygen, nitrogen, argon, hydrogen, deuterium, carbon monoxide, and carbon dioxide.
The set contains all items needed for immediate use:
The set contains all items needed for immediate use:
- 316 stainless steel adapter and matched PTFE sleeve
- 1 pressure gauge, 300 psi, 2 valves
- 1 pressure tube, 35 mL
- 20 septa
- All fittings for connecting to a gas regulator
Legal Information
Q-Tube is a trademark of Q Labtech LLC