Z740596
Aldrich® Essentials beaker, Berzelius
tall form, capacity 50 mL, volume 10-40 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
feature
subdivision 10 mL
packaging
pkg of 12 ea
manufacturer/tradename
Sigma-Aldrich
volume measuring range
10-40 mL
capacity
50 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Borosilicate glass, Berzelius, tall form beaker with reference graduations and pour spout. Standard uniform walls and flat bottom for even heat distribution.
Other Notes
Browse all glass beakers
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service