Key Documents
Z645753
Chemical spill kit bag
Synonym(s):
chemical cleanup, spill cleanup, spill safety
Select a Size
1 KIT
₹39,703.20
₹39,703.20
Available to ship on19 April 2025Details
Select a Size
Change View
1 KIT
₹39,703.20
About This Item
UNSPSC Code:
24111500
NACRES:
NB.35
₹39,703.20
Available to ship on19 April 2025Details
manufacturer/tradename
Clear-spill CK3
availability
not available in ROW, USA and Canada
General description
Kit designed for small to medium spills, ideal for laboratories or stores. Contains 40 pads, 4 x 8 cm by 1.2 m socks, 1 x 8 cm by 3 m sock, 1 x 30 cm by 35 cm cushion, 1 x 35 cm by 45 cm cushion with instructions supplied in a waterproof zip top bag with carry handles. Kit will absorb 57 litres.
PLEASE NOTE: KIT NO LONGER CONTAINS PPE
Choose from one of the most recent versions:
Certificates of Analysis (COA)