Z620157
DURAN disposable culture tubes
H × diam. 75 mm × 12.25 mm
Synonym(s):
231720965
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
AR-Glas®
clear tube
round bottom tube
packaging
pack of 500
manufacturer/tradename
DURAN®
H × diam.
75 mm × 12.25 mm
capacity
7 mL
wall size
0.55 mm
Looking for similar products? Visit Product Comparison Guide
General description
with straight rim
Legal Information
AR-Glas is a registered trademark of Schott AG
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service