Documents
Z560758
Sigma-Aldrich
Scienceware® fitting assortment kit
28 fittings to handle the most common laboratory connections
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F19626-0000
General description
28 fittings to handle the most common laboratory connections. Fittings can handle teperatures up to 135°C.
Contains:
  • 5 Straight connectors
  • 3 Stepped fittings
  • 8 Bifurcated ("Y") fittings
  • 5 "T" connectors
  • 5 Quick-disconnect fittings
  • 2 Tubing clamps
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP