Key Documents
Z551511
Torion screw threaded plastic plug
#14
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
description
For 14 mm tubing
packaging
pkg of 5 ea
General description
#8 will fit a 5/16in. O.D. size, and #14 will fit a 1/2in. O.D. size, and 3/16in. O.D. fits a #5 Torion.
Legal Information
Torion is a trademark of Bibby Sterlin Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)