Key Documents
Z258660
Sigma-Aldrich
Atlas of Organic Conversion Chart: An Aid in Perspective
set contains a manual and two large colorful charts
About This Item
UNSPSC Code:
60104506
L × W
45 in. × 34 in. , chart dimensions
General description
These charts are designed to show the connectivity between preparations and chemical properties which constitute organic conversions and highlight multi-step strategies for synthesizing simple molecules. This adds a new dimension to the teaching and learning of organic chemistry and helps minimize difficulties in solving problems related to functional group transformations.
Part I
Aliphatic compounds: contains 595 acyclic and alicyclic structures (including some simple heterocycles). Structures and reagents are represented in black, and a particular class of compounds is found in the specified portion of the chart.
Part II
Aromatic compounds: contains 495 aromatic structures (benzene and its derivatives). Structures are also drawn in black while heteroatom functional groups are red for immediate perception; each class of compounds is spread out but still remains connected.
Solvents in both charts are indicated in blue, and catalysts are blue with a red asterisk. Set contains a manual and two large colorful charts, dim.: L x W 45 in. × 34 in.
Choose from one of the most recent versions:
Certificates of Analysis (COA)