Documents
Z189855
Sigma-Aldrich
Spinplus® magnetic stirring bar
size 3/4 in. × 3/4 in.
Synonym(s):
Stir Bar, Stirbar, Stirring Bar
About This Item
UNSPSC Code:
41103806
eCl@ss:
32100100
NACRES:
NB.72
material
PTFE-coated
cross PTFE-coated bar
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37144-0034
L
19.1 mm
diam.
9.5 mm
size
3/4 in. × 3/4 in.
General description
Cross-shaped bars create a deep vortex for stable operations.
Legal Information
Spinplus is a registered trademark of SP Scienceware, a division of SP