Documents
Z116653
Sigma-Aldrich
Aldrich® gas-condensing bottle
with hose connection
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
150 mL
Related Categories
General description
With stopcock, graduated, holds 150 mL of liquefied gas.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC