Skip to Content
Merck
All Photos(1)

Key Documents

W506605

Sigma-Aldrich

Methyl (p-tolyloxy)acetate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OCH2CO2CH3
CAS Number:
Molecular Weight:
180.20
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

Quality Level

grade

Kosher

Assay

≥98%

bp

119 °C/5 mmHg (lit.)

application(s)

flavors and fragrances

Organoleptic

fruity

SMILES string

COC(=O)COc1ccc(C)cc1

InChI

1S/C10H12O3/c1-8-3-5-9(6-4-8)13-7-10(11)12-2/h3-6H,7H2,1-2H3

InChI key

BUKHBVPVKOXGSO-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service