Skip to Content
Merck
All Photos(1)

Documents

W500607

Sigma-Aldrich

Citronellyl tiglate, mixture of isomers

≥96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)CO2CH2CH2CH(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
238.37
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.340

biological source

synthetic

grade

Kosher

reg. compliance

FDA 21 CFR 117

Assay

≥96%

may contain

<0.10% alpha-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.465 (lit.)

bp

144-145 °C/9 mmHg (lit.)

density

0.903 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; rose

SMILES string

CCC(=C)C(=O)OCCC(C)CC\C=C(\C)C.C\C=C(/C)C(=O)OCCC(C)CC\C=C(\C)C

InChI

1S/2C15H26O2/c2*1-6-14(5)15(16)17-11-10-13(4)9-7-8-12(2)3/h6,8,13H,7,9-11H2,1-5H3;8,13H,5-7,9-11H2,1-4H3/b14-6+;

InChI key

WWYVFYUOASWQQR-JSSTZBRYSA-N

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service