Skip to Content
Merck
All Photos(2)

Documents

W375420

Sigma-Aldrich

Vanillin isobutyrate

natural, ≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2C6H3-2-(OCH3)-4-(CHO)
CAS Number:
Molecular Weight:
222.24
FEMA Number:
3754
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.811

biological source

botanical

grade

FG
Fragrance grade
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

refractive index

n20/D 1.524 (lit.)

density

1.12 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

creamy; nutmeg; fruity; sweet; vanilla

SMILES string

COc1cc(C=O)ccc1OC(=O)C(C)C

InChI

1S/C12H14O4/c1-8(2)12(14)16-10-5-4-9(7-13)6-11(10)15-3/h4-8H,1-3H3

InChI key

BGKAKRUFBSTALK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

334.4 °F

Flash Point(C)

168 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service