Skip to Content
Merck
All Photos(1)

Key Documents

W305805

Sigma-Aldrich

(+/-)-Tetrahydrofurfuryl propionate

≥96%

Synonym(s):

DL-Tetrahydrofurfuryl propionate, N-Propionic acid tetrahydrofurfuryl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O3
CAS Number:
Molecular Weight:
158.19
FEMA Number:
3058
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.049

biological source

synthetic

grade

Halal
Kosher

Assay

≥96%

refractive index

n20/D 1.438 (lit.)

bp

207 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

violet

SMILES string

CCC(=O)OCC1CCCO1

InChI

1S/C8H14O3/c1-2-8(9)11-6-7-4-3-5-10-7/h7H,2-6H2,1H3

InChI key

FMKCDSXOYLTWBR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service