Skip to Content
Merck
All Photos(1)

Key Documents

W268607

Sigma-Aldrich

α-Methylbenzyl butyrate

≥98%, FG

Synonym(s):

Styrallyl butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH(CH3)C6H5
CAS Number:
Molecular Weight:
192.25
FEMA Number:
2686
EC Number:
Council of Europe no.:
2083
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.231
NACRES:
NA.21

biological source

synthetic

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117

Assay

≥98%

refractive index

n20/D 1.487 (lit.)

bp

83-84 °C/3 mmHg (lit.)

density

0.99 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; fruity; earthy; sweet

SMILES string

CCCC(=O)OC(C)c1ccccc1

InChI

1S/C12H16O2/c1-3-7-12(13)14-10(2)11-8-5-4-6-9-11/h4-6,8-10H,3,7H2,1-2H3

InChI key

GGKADXREVJTZMF-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service