Skip to Content
Merck
All Photos(1)

Documents

W255203

Sigma-Aldrich

Heptyl formate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2(CH2)6CH3
CAS Number:
Molecular Weight:
144.21
FEMA Number:
2552
EC Number:
Council of Europe no.:
341
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.074

biological source

synthetic

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.413 (lit.)

bp

178 °C (lit.)

density

0.882 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

apple; fatty; plum

SMILES string

[H]C(=O)OCCCCCCC

InChI

1S/C8H16O2/c1-2-3-4-5-6-7-10-8-9/h8H,2-7H2,1H3

InChI key

XEAMDSXSXYAICO-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Feng et al.
Journal of food science, 85(10), 3273-3281 (2020-09-13)
Huangjiu is a traditional wine in China with special taste and flavor. However, changes of aroma compounds during storage of Huangjiu remain unclear. In this study, aroma compounds in Jinse Nianhua Huangjiu of three different storage ages were qualitatively and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service