Skip to Content
Merck
All Photos(1)

Key Documents

W221406

Sigma-Aldrich

Butyl stearate

≥95%

Synonym(s):

Stearic acid butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COO(CH2)3CH3
CAS Number:
Molecular Weight:
340.58
FEMA Number:
2214
Beilstein:
1792866
EC Number:
Council of Europe no.:
2189c
MDL number:
Flavis number:
9.246
Pricing and availability is not currently available.

Assay

≥95%

composition

butyl palmitate, ≥40%
butyl stearate, ≥40%

density

0.861 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCCCC

InChI

1S/C22H44O2/c1-3-5-7-8-9-10-11-12-13-14-15-16-17-18-19-20-22(23)24-21-6-4-2/h3-21H2,1-2H3

InChI key

ULBTUVJTXULMLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Slightly fatty odor

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service