Skip to Content
Merck
All Photos(1)

Key Documents

W221228

Sigma-Aldrich

Isobutyl propionate

natural, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
130.18
FEMA Number:
2212
EC Number:
Council of Europe no.:
406c
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.125
Pricing and availability is not currently available.

grade

Kosher
natural

Assay

≥98%

refractive index

n20/D 1.397 (lit.)

bp

66 °C/60 mmHg (lit.)

mp

−71 °C (lit.)

density

0.869 g/mL at 25 °C (lit.)

SMILES string

CCC(=O)OCC(C)C

InChI

1S/C7H14O2/c1-4-7(8)9-5-6(2)3/h6H,4-5H2,1-3H3

InChI key

FZXRXKLUIMKDEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Heavy floral

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service