Skip to Content
Merck
All Photos(1)

Key Documents

W208108

Sigma-Aldrich

Isoamyl phenylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
FEMA Number:
2081
EC Number:
Council of Europe no.:
2161
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.789
Pricing and availability is not currently available.

refractive index

n20/D 1.485 (lit.)

bp

268 °C (lit.)

density

0.975 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCOC(=O)Cc1ccccc1

InChI

1S/C13H18O2/c1-11(2)8-9-15-13(14)10-12-6-4-3-5-7-12/h3-7,11H,8-10H2,1-2H3

InChI key

QWBQBUWZZBUFHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural Occurrence: Peppermint oil.

Biochem/physiol Actions

Odor at 1%
Taste at 5-10 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

217.9 °F

Flash Point(C)

103.3 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 25, 71-71 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service