Key Documents
TEM-FFT300CU25
Sigma-Aldrich
Formvar Supported Copper Thin Bar Square Mesh
size 300 mesh, box of 25
Synonym(s):
FORMVAR SUPPORT FILM THIN BAR SQUARE 300 MESH CU GRIDS
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-FFT300CU25 is not currently sold in your country. Contact Technical Service
packaging
box of 25
size
300 mesh
General description
A 10 nm thin film of pure formvar resin supported thin bar square mesh grids. Copper grid, 300 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)