Documents
SYNF461025
Synthware storage flask with standard taper side arm
capacity 25 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonym(s):
Synthware F461025, round flask with side arm
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F461025
capacity
25 mL
valve size
0-4 mm
General description
Supplied with high vacuum valve with PTFE O-ring at tip.
Legal Information
Synthware is a trademark of Kemtech America Inc.