Documents
SYNF459100
Synthware Squibb-style separatory funnel with standard taper joints, PTFE stopcock and PTFE stopper
100 mL, joint: ST/NS 19/22, PTFE stopcock and PTFE stopper
Synonym(s):
Synthware F459100, glass separatory funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F459100
capacity
100 mL
100 mL
Related Categories
General description
With 2 mm PTFE stopcock. Supplied with a glass stopper or PTFE stopper.
Legal Information
Synthware is a trademark of Kemtech America Inc.