Skip to Content
Merck
All Photos(1)

Key Documents

N19001

Sigma-Aldrich

3-Nitro-1,8-naphthalic anhydride

technical grade

Synonym(s):

3-Nitronaphthalene-1,8-dicarboxylic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H5NO5
CAS Number:
Molecular Weight:
243.17
Beilstein:
23762
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
To inquire about this product N19001, please contact your local Merck office or dealer. Contact Technical Service

grade

technical grade

mp

247-249 °C (dec.) (lit.)

SMILES string

[O-][N+](=O)c1cc2C(=O)OC(=O)c3cccc(c1)c23

InChI

1S/C12H5NO5/c14-11-8-3-1-2-6-4-7(13(16)17)5-9(10(6)8)12(15)18-11/h1-5H

InChI key

FLFLZYYDLIKGJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service