Skip to Content
Merck
All Photos(1)

Key Documents

H3145

Sigma-Aldrich

Sodium (S)-3-hydroxybutyrate

~97%

Synonym(s):

(S)-3-Hydroxybutanoic acid sodium salt, (S)-3-Hydroxybutyric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NaO3
CAS Number:
Molecular Weight:
126.09
Beilstein:
1720567
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

~97%

storage temp.

2-8°C

SMILES string

[Na+].C[C@H](O)CC([O-])=O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service