Documents
DWK73750-20125
KIMBLE® disposable screw thread culture tube with white marking spot
round bottom borosilicate glass, tube diam. × L (20 mm) (125 mm), non-sterile
About This Item
UNSPSC Code:
41121706
NACRES:
NB.11
material
round bottom borosilicate glass
round bottom
screw top neck
transparent borosilicate glass tube
sterility
non-sterile
feature
without cap
packaging
case of 500 ea Modular tray shrink-wrapped in plastic film
manufacturer/tradename
Kimble® 73750-20125
tube diam. × L
(20 mm) (125 mm)
tube diam.
(20 mm)
fitting
neck (External Thread)
General description
Disposable Screw Thread Culture Tubes with Marking Spot
  • These culture tubes are suitable for tissue culture work and general bacteriological use.
  • Round-bottomed
  • Each tube has a chevron marking area
  • Supplied without caps
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Features and Benefits
• Manufactured from borosilicate glass 5.1• Round-bottomed• Each tube has a chevron marking area• Modular trays are shrink-wrapped in plastic film• Supplied without caps
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences