D143359
Dansyl chloride
98%
Synonym(s):
5-(Dimethylamino)naphthalene-1-sulfonyl chloride, DNSCl
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H12ClNO2S
CAS Number:
Molecular Weight:
269.75
Beilstein:
2217205
EC Number:
MDL number:
UNSPSC Code:
12352100
Recommended Products
Assay
98%
mp
72-74 °C (lit.)
SMILES string
CN(C)c1cccc2c(cccc12)S(Cl)(=O)=O
InChI
1S/C12H12ClNO2S/c1-14(2)11-7-3-6-10-9(11)5-4-8-12(10)17(13,15)16/h3-8H,1-2H3
InChI key
XPDXVDYUQZHFPV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
A fluorogenic reagent for fluorescent N-terminal derivatization of amino acids and peptides and detection by reverse phase HPLC.
Reagent for fluorescent labeling of amines, amino acids, proteins, and phenols.
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chemical & Pharmaceutical Bulletin, 37, 2841-2841 (1989)
Cristina Avonto et al.
Toxicology and applied pharmacology, 289(2), 177-184 (2015-10-13)
Skin sensitization is an important toxicological end-point in the risk assessment of chemical allergens. Because of the complexity of the biological mechanisms associated with skin sensitization, integrated approaches combining different chemical, biological and in silico methods are recommended to replace
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service