Skip to Content
Merck
All Photos(1)

Key Documents

CDS021725

Sigma-Aldrich

1-(4-Fluorophenyl)propan-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12FN
Molecular Weight:
153.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

SMILES string

CC(CC1=CC=C(C=C1)F)N

InChI

1S/C9H12FN/c1-7(11)6-8-2-4-9(10)5-3-8/h2-5,7H,6,11H2,1H3

InChI key

DGXWNDGLEOIEGT-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

13 - Non Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Zwartsen et al.
Toxicology, 422, 69-75 (2019-04-23)
Many psychoactive substances affect the human dopamine (DA) reuptake transporter (hDAT). Polymorphisms in the encoding gene could affect the functionality of the transporter and consequently alter effects of psychotropic and recreational drugs. Recently, a T356 M single nucleotide polymorphism in the
Sebastiaan F Teunissen et al.
Journal of the American Society for Mass Spectrometry, 28(12), 2665-2676 (2017-09-08)
Paper spray tandem mass spectrometry is used to identify and quantify eight individual amphetamines in whole blood in 1.3 min. The method has been optimized and fully validated according to forensic toxicology guidelines, for the quantification of amphetamine, methamphetamine, 3,4-methylenedioxyamphetamine
Enrico Gerace et al.
Journal of separation science, 42(8), 1577-1584 (2019-02-12)
Most routine practices for drugs-of-abuse testing do not include screening procedures for new psychoactive substances, despite their increasing diffusion, preventing clear knowledge of the real consumption of these drugs in the populations. To make up for this shortcoming, a gas
Xun Zhou et al.
Cells, 9(4) (2020-04-17)
Hyperthermia is one of the severe acute adverse effects that can be caused by the ingestion of recreational drugs, such as methcathinones. The effect of hyperthermia on neurotoxicity is currently not known. The primary aim of our study was therefore
Xun Zhou et al.
International journal of molecular sciences, 21(8) (2020-04-25)
Halogenation of amphetamines and methcathinones has become a common method to obtain novel psychoactive substances (NPS) also called "legal highs". The para-halogenated derivatives of amphetamine and methcathinone are available over the internet and have entered the illicit drug market but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service