Skip to Content
Merck
All Photos(1)

Key Documents

C72900

Sigma-Aldrich

5-Chlorovaleric acid

98%

Synonym(s):

5-Chloropentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)4COOH
CAS Number:
Molecular Weight:
136.58
Beilstein:
1745183
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.454 (lit.)

mp

18 °C (lit.)

density

1.170 g/mL at 20 °C (lit.)

SMILES string

OC(=O)CCCCCl

InChI

1S/C5H9ClO2/c6-4-2-1-3-5(7)8/h1-4H2,(H,7,8)

InChI key

YSXDKDWNIPOSMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service