Skip to Content
Merck
All Photos(1)

Documents

BR781671

BRAND® microplate BRANDplates®, pureGrade

size 96 wells, size 330 μL, flat bottom, black, with transparent bottom

Synonym(s):

BRANDplates®, Microplates, pureGradeS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

black wells
flat bottom
polystyrene

sterility

sterile

feature

lid
skirt (F-bottom)
with lid

packaging

pack of 1 ea
pkg of (individually packaged)

manufacturer/tradename

BRAND 781671

size

330 μL
96 wells

working volume

330 μL

color

black, with transparent bottom

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® Microplate BRANDplates® PureGrade features a non-treated surface, making it particularly well-suited for bacteriological assays. Sterile according to ISO 11137 standards, this microplate ensures a contamination-free environment for sensitive experiments. It is classified as BIO-CERT® CELL CULTURE QUALITY, guaranteeing that it is free from human DNA, RNase, DNase, and pyrogens, and is non-cytotoxic. This reliable microplate is an essential tool for researchers conducting microbiological studies, providing consistent performance and safety in cell culture applications.

Features and Benefits

• Optimal surfaces for optimal results• Reliable quality from the cleanroom• Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service