Skip to Content
Merck
All Photos(1)

Documents

BR126963

BRAND® reservoir bottle

volume 1000 mL, without dosing head

Synonym(s):

Accessories, Bottles, Dispensers, tilt measure, Reservoir bottles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.01

material

clear borosilicate glass
colorless
narrow-mouth bottle
round bottle

description

NS 29/32 DIN 12 463

sterility

non-sterile

feature

cap: no

packaging

pack of 1 ea

manufacturer/tradename

BRAND 126963

capacity

1000 mL

volume

1000 mL

General description

The BRAND® Reservoir Bottle is crafted from high-quality soda-lime glass, offering excellent durability and chemical resistance for laboratory use. With a generous capacity of 1000 ml, this bottle is designed to accommodate a variety of liquids and solutions. It features a neck size of 29/32, making it compatible with a range of laboratory accessories and closures. This reservoir bottle is ideal for storing, mixing, and dispensing liquids, making it an essential tool for researchers and technicians in various scientific applications.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service