Skip to Content
Merck
All Photos(1)

Documents

BMSR2020500

Benchmark RotoMini Rotator Tube holder

to hold, 6 x 50 mL

Synonym(s):

tube rotator, mini tube rotator, lab roller, mini lab roller, lab revolver, rotoflex, rotoflex, argos rotator, mini rotator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804
NACRES:
NB.11

feature

to hold 6 x 50 mL

manufacturer/tradename

Benchmark Scientific R2020-500

Looking for similar products? Visit Product Comparison Guide

Application

Mixing of samples in tubes

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service