Skip to Content
Merck
All Photos(1)

Key Documents

B12253

Sigma-Aldrich

Nα-Benzoyl-L-arginine ethyl ester hydrochloride

99%

Synonym(s):

BAEE, BAEE hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NH(CH2)3CH(NHCOC6H5)CO2C2H5·HCl
CAS Number:
Molecular Weight:
342.82
Beilstein:
3781694
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

99%

optical activity

[α]22/D −15.5°, c = 2 in 1 M HCl

mp

127-131 °C (lit.)

SMILES string

Cl.CCOC(=O)[C@H](CCCNC(N)=N)NC(=O)c1ccccc1

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service