Skip to Content
Merck
All Photos(1)

Key Documents

938920

Sigma-Aldrich

4′-Hydroxy-4,5-dimethoxy-[1,1′-biphenyl]-2-carbonitrile

Synonym(s):

Maiti-Maji Auxiliary

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO3
CAS Number:
Molecular Weight:
255.27
UNSPSC Code:
12352142
Pricing and availability is not currently available.

form

powder

Quality Level

color

light beige

functional group

nitrile

storage temp.

−20°C

SMILES string

COC1=C(OC)C=C(C2=CC=C(O)C=C2)C(C#N)=C1

InChI key

LMDUSGRXYIRJDS-UHFFFAOYSA-N

General description

4′-Hydroxy-4,5-dimethoxy-[1,1′-biphenyl]-2-carbonitrile is an effective temporary directing group (TDG) for meta directed C-H functionalization, effective in many transformations including olefination, silylation, and cyanation of arenes.

Application

A temporary directing group (TDG) to assist as a co-catalyst for metal catalyzed C-H functionalization. Often in C-H functionalization, an auxiliary compound is used to control site selectivity. These traditionally are covalently bonded to the compound of interest, and must subsequently be removed after functionalization, like a typical protecting group. To simplify the process of C-H functionalization, 2-fluoro-6-(pyrimidin-5-yl)aniline is one of a series of temporary directing groups developed by Deb Maiti′s lab that promote site selectivity without the inclusion of additional synthetic steps.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H-bonded reusable template assisted para-selective ketonisation using soft electrophilic vinyl ethers
Maji, Arun et. al.
Nature Communications, 9(1) (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service