Skip to Content
Merck
All Photos(1)

Key Documents

92409

Sigma-Aldrich

1-Butyl-1-methylpyrrolidinium tetrafluoroborate

≥97.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20BF4N
CAS Number:
Molecular Weight:
229.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
crystals
Assay:
≥97.0% (T)

Assay

≥97.0% (T)

form

crystals

impurities

≤0.2% halogens (as chloride)
≤0.5% water

SMILES string

F[B-](F)(F)F.CCCC[N+]1(C)CCCC1

InChI

1S/C9H20N.BF4/c1-3-4-7-10(2)8-5-6-9-10;2-1(3,4)5/h3-9H2,1-2H3;/q+1;-1

InChI key

PGCVCJOPLBWQHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Butyl-1-methylpyrrolidinium tetrafluoroborate ([BMPY][BF4]) can be used:
  • As an electrolyte along with 1,2-butylene carbonate and 3-cyanopropionic acid methyl ester solvents, applicable in the electrochemical double-layer capacitors (EDLCs).[1][2]
  • As an ionic solvent in the N-alkylation reaction of acridine ester precursors with 1,3-propane sultone.[3]
  • As an inhibitor of corrosion for copper in acidic conditions.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 2-butylene carbonate as solvent for EDLCs
Hess LH and Balducci A
Electrochimica Acta, 281, 437-444 (2018)
Facile N-alkylation of acridine esters with 1, 3-propane sultone in ionic liquids
Natrajan A and Wen D
Green Chemistry, 13(4), 913-921 (2011)
Cyano ester as solvent for high voltage electrochemical double layer capacitors
Schutter C, et al.
Electrochimica Acta, 224, 278-284 (2017)
Impact of some pyrrolidinium ionic liquids on copper dissolution behavior in acidic environment: experimental, morphological and theoretical insights
El-Katori EE and Abousalem AS
Royal Society of Chemistry Advances, 9(36), 20760-20777 (2019)
Susana P F Costa et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 18(10), 1351-1357 (2017-02-22)
A novel automated fluorimetric technique was developed for the assessment of the chemical oxygen demand (COD) of ionic liquids (ILs) and combined with a photodegradation step to promote IL degradation. The method was implemented on a sequential injection analysis (SIA)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service