Skip to Content
Merck
All Photos(1)

Documents

907901

Sigma-Aldrich

(2S,3R)-3-(Benzyloxy)-2-((3-(tert-butyl)-2-hydroxybenzyl)amino)-N,N-dimethylbutanamide

≥95%

Synonym(s):

Hoveyda aminophenol catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34N2O3
Molecular Weight:
398.54
UNSPSC Code:
12352111

Assay

≥95%

form

paste

storage temp.

2-8°C

Application

(2S,3R)-3-(Benzyloxy)-2-((3-(tert-butyl)-2-hydroxybenzyl)amino)-N,N-dimethylbutanamide is an aminophenol organocatalyst developed by the Hoveyda lab for enantioselective addition of allylboron compounds to imines, ketones, and other carbonyls, including polyfluorinated substrates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service