Skip to Content
Merck
All Photos(1)

Key Documents

907332

Sigma-Aldrich

3-(3-Methyl-3H-diazirin-3-yl)-N-(3-(triethoxysilyl)propyl)propanamide

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H29N3O4Si
Molecular Weight:
331.48
UNSPSC Code:
12352200

Assay

≥95%

form

liquid

reaction suitability

reagent type: cross-linking reagent

availability

available only in USA

storage temp.

−20°C

Application

3-(3-Methyl-3H-diazirin-3-yl)-N-(3-(triethoxysilyl)propyl)propanamide is a photoinducible silane diazirine useful for the immobilization of protein targets onto surfaces. Upon UV light (~360 nm) irradiation, a covalent bond is formed with nearby protein residues, binding it to the surface. Ye, et a,l demonstrated its use in immobilizing antibodies for development of a silica-based fiber optic biosensor (FOB).

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service