Skip to Content
Merck
All Photos(2)

Key Documents

900268

Sigma-Aldrich

Bis(3,5-bis(trifluoromethyl)phenyl)(2′,6′-bis(isopropoxy)-3,6-dimethoxybiphenyl-2-yl)phosphine

≥95%

Synonym(s):

2-{Bis[3,5-bis(trifluoromethyl)phenyl]phosphino}-3,6-dimethoxy-2′,6′-bis(isopropoxy)-1,1′-biphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H31F12O4P
CAS Number:
Molecular Weight:
786.58
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reagent type: ligand
reaction type: Cross Couplings

functional group

phosphine

Application

Bis(3,5-bis(trifluoromethyl)phenyl)(2′,6′-bis(isopropoxy)-3,6-dimethoxybiphenyl-2-yl)phosphine is the Pd G4 complex from the Buchwald</A group, offers best-in-class performance in the arylation of sterically hindered secondary amines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathaniel H Park et al.
Angewandte Chemie (International ed. in English), 54(28), 8259-8262 (2015-06-03)
In Pd-catalyzed C-N cross-coupling reactions, α-branched secondary amines are difficult coupling partners and the desired products are often produced in low yields. In order to provide a robust method for accessing N-aryl α-branched tertiary amines, new catalysts have been designed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service