Skip to Content
Merck
All Photos(1)

Key Documents

855413

Sigma-Aldrich

1-Naphthyl phosphate monosodium salt monohydrate

98%

Synonym(s):

α-Naphthyl phosphate monosodium salt monohydrate, Sodium 1-naphthyl phosphate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OP(O)(OH)(ONa) · H2O
CAS Number:
Molecular Weight:
264.15
Beilstein:
1877764
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

98%

SMILES string

[Na+].[H]O[H].OP([O-])(=O)Oc1cccc2ccccc12

InChI

1S/C10H9O4P.Na.H2O/c11-15(12,13)14-10-7-3-5-8-4-1-2-6-9(8)10;;/h1-7H,(H2,11,12,13);;1H2/q;+1;/p-1

InChI key

NSWUDGODQXQNON-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Preferred substrate for bioanalytical determinations of acid phosphatases, such as prostatic acid phosphatase.

Biochem/physiol Actions

Non-specific phosphatase inhibitor. Inhibits acid, alkaline and protein phosphatases.

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service