Skip to Content
Merck
All Photos(1)

Documents

808040

Sigma-Aldrich

Copper

nanowires, diam. × L ~100 nm (±20 nm) × 10~20 μm, Wet cake (Wet cake)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
Molecular Weight:
63.55
UNSPSC Code:
12141711
NACRES:
NA.23

Quality Level

form

nanowires

diam. × L

~100 nm (±20 nm) × 10~20 μm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Copper nanowires find applications in flexible electronics, transparent electrodes, photocatalysis, solar cells and as shape memory material [1, 2, 3].

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service