Skip to Content
Merck
All Photos(1)

Key Documents

79520

Sigma-Aldrich

Trimethylsilyl P,P-diethylphosphonoacetate

Synonym(s):

Diethyl (trimethylsilyloxycarbonylmethyl)phosphonate, Phosphonoacetic acid P,P-diethyl trimethylsilyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(O)CH2CO2Si(CH3)3
CAS Number:
Molecular Weight:
268.32
Beilstein:
2371028
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
Pricing and availability is not currently available.

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.430 (lit.)

bp

93 °C/0.05 mmHg

density

1.059 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOP(=O)(CC(=O)O[Si](C)(C)C)OCC

InChI

1S/C9H21O5PSi/c1-6-12-15(11,13-7-2)8-9(10)14-16(3,4)5/h6-8H2,1-5H3

InChI key

VHIDZGBMGSRVHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A Horner-Wittig reagent for the preparation of α,β-unsaturated acids[1][2]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L. Lombardo et al.
Synthesis, 131-131 (1978)
Synthetic Communications, 8, 463-463 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service