Skip to Content
Merck
All Photos(1)

Documents

794368

Sigma-Aldrich

3-(Trimethoxysilyl)propyl vinyl carbamate

contains ≤500 ppm phenothiazine as inhibitor, 95%

Synonym(s):

Vinyl [3-(trimethoxysilyl)propyl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NO5Si
CAS Number:
Molecular Weight:
249.34
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

95%

form

liquid

contains

≤500 ppm phenothiazine as inhibitor

density

1.124 g/mL at 25 °C

storage temp.

−20°C

SMILES string

C=COC(NCCC[Si](OC)(OC)OC)=O

InChI

1S/C9H19NO5Si/c1-5-15-9(11)10-7-6-8-16(12-2,13-3)14-4/h5H,1,6-8H2,2-4H3,(H,10,11)

InChI key

ABJBSCWBVKPQQP-UHFFFAOYSA-N

Application

This monomer is commonly used in the formulation of hydrogel for contact lenses and other ophthalmic devices.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service