Skip to Content
Merck
All Photos(1)

Key Documents

767085

Sigma-Aldrich

2-bromo-3,4,5-trimethoxy-benzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11BrO4
CAS Number:
Molecular Weight:
275.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

67-73 °C

functional group

aldehyde
bromo

SMILES string

COc1cc(C=O)c(Br)c(OC)c1OC

InChI

1S/C10H11BrO4/c1-13-7-4-6(5-12)8(11)10(15-3)9(7)14-2/h4-5H,1-3H3

InChI key

LADPQZFHJVANIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service