Skip to Content
Merck
All Photos(1)

Key Documents

766194

Sigma-Aldrich

1-Boc-2-phenylhydrazine

97%

Synonym(s):

1-(tert-Butoxycarbonyl)-2-phenylhydrazine, 2-Phenylhydrazinecarboxylic acid 1,1-dimethylethyl ester, 3-Phenylcarbazic acid tert-butyl ester, N-Boc-N′-phenylhydrazine, tert-Butyl-2-phenylhydrazinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O2
CAS Number:
Molecular Weight:
208.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

90-94 °C

functional group

amine
hydrazide

SMILES string

CC(C)(C)OC(=O)NNc1ccccc1

InChI

1S/C11H16N2O2/c1-11(2,3)15-10(14)13-12-9-7-5-4-6-8-9/h4-8,12H,1-3H3,(H,13,14)

InChI key

PYEQNXVGJRAEIG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service