Skip to Content
Merck
All Photos(1)

Key Documents

745634

Sigma-Aldrich

3-Bromo-4-(difluoromethyl)pyridine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BrF2N
CAS Number:
Molecular Weight:
208.00
UNSPSC Code:
12352100
NACRES:
NA.22

Assay

95%

form

liquid

storage temp.

2-8°C

InChI

1S/C6H4BrF2N/c7-5-3-10-2-1-4(5)6(8)9/h1-3,6H

InChI key

KCMMLUVZVBBEFO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service