74399
Cysteamine 2-chlorotrityl thioether polymer-bound
extent of labeling: 0.3-1 mmol/g loading
Synonym(s):
2-Aminoethyl 2-chlorotrityl thioether polymer-bound, S-(2-Chlorotrityl)cysteamine polymer-bound
About This Item
extent of labeling
0.3-1 mmol/g loading
matrix
crosslinked with 1% DVB
particle size
200-400 mesh
storage temp.
2-8°C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service