Skip to Content
Merck
All Photos(2)

Key Documents

741264

Sigma-Aldrich

Potassium 2-fluoropyrimidine-5-trifluoroborate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2BF4KN2
Molecular Weight:
203.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

321-326 °C

functional group

fluoro

SMILES string

[K+].Fc1ncc(cn1)[B-](F)(F)F

InChI

1S/C4H2BF4N2.K/c6-4-10-1-3(2-11-4)5(7,8)9;/h1-2H;/q-1;+1

InChI key

NCUUVZSLKMYISK-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service