Skip to Content
Merck
All Photos(1)

Key Documents

737119

Sigma-Aldrich

2,2-difluoro-1,3-dimethylimidazolidine

95%

Synonym(s):

DFI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10F2N2
CAS Number:
Molecular Weight:
136.14
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.402

density

1.096 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CN1CCN(C)C1(F)F

InChI

1S/C5H10F2N2/c1-8-3-4-9(2)5(8,6)7/h3-4H2,1-2H3

InChI key

MGDCBOKBTJIJBT-UHFFFAOYSA-N

Application

A nucleophilic fluorinating agent.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.9 °F

Flash Point(C)

33.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angelika Lehner et al.
BMC microbiology, 6, 15-15 (2006-03-01)
Enterobacter sakazakii is a foodborne pathogen that has been associated with sporadic cases and outbreaks causing meningitis, necrotizing enterocolitis and sepsis especially in neonates. The current FDA detection method includes two enrichment steps, the subculturing of the second enrichment broth
Shaomin Chen et al.
Diagnostic pathology, 7, 40-40 (2012-04-17)
The amplification of oncogenes initiated by high-risk human papillomavirus (HPV) infection is an early event in cervical carcinogenesis and can be used for cervical lesion diagnosis. We measured the genomic amplification rates and the patterns of human telomerase RNA gene
Marisol J Voncken et al.
Journal of behavior therapy and experimental psychiatry, 69, 101574-101574 (2020-05-30)
This study aimed to unravel the relationship between socially anxious individuals' expectation of being (dis)liked and actual likeability by looking at the mediating role of both strategic and automatic social behavior: Self-disclosure as well as mimicry were examined. Female participants
Walid M El-Sharoud et al.
BMC microbiology, 9, 24-24 (2009-02-04)
Cronobacter is a recently proposed genus consisting of six genomospecies that encompass the organisms previously identified as Enterobacter sakazakii. Cronobacter are opportunistic pathogens and are known to cause serious infections in infants, particularly neonates. High case fatality rates have been
Joanna Jurewicz et al.
Systems biology in reproductive medicine, 61(1), 37-43 (2014-11-08)
The present study was designed to investigate whether environmental exposure to pyrethroids was associated with sperm DNA damage. Between January 2008 and April 2011 286 men under 45 years of age with a normal sperm concentration of 15-300 10(6)/ml [WHO 2010]

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service