Skip to Content
Merck
All Photos(1)

Key Documents

728616

Sigma-Aldrich

4-(Trifluoroacetyl)benzoyl chloride

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4ClF3O2
CAS Number:
Molecular Weight:
236.58
Beilstein:
2728981
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (GC)
97.0-103.0% (T)

form

liquid

SMILES string

FC(F)(F)C(=O)c1ccc(cc1)C(Cl)=O

InChI

1S/C9H4ClF3O2/c10-8(15)6-3-1-5(2-4-6)7(14)9(11,12)13/h1-4H

InChI key

RBOMQQAMVJDXBU-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

249.1 °F

Flash Point(C)

120.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service